Thymic Factor Trifluoroacetate
Supplier: BACHEM AMERICAS INC
4009914.0005
4009914.0025
H-4865.0005BAEA
387.21
CAD
H-4865.0005BA
H-4865.0025BA
Thymic Factor Trifluoroacetate
Proteins and Peptides
See also the product families 'Adjuvant Peptides' and 'Thymosins and Fragments'.
For exerting its anti-inflammatory and immunomodulatory activity, the peptide hormone thymulin, pEAKSQGGSN, requires the presence of Zn(II). Thymulin showed promising results in experimental models of lung diseases as well as potential as an analgesic peptide.