Human pTH (1-34) Acetate
Supplier: BACHEM AMERICAS INC
4011474.0001 4011474.0005
H-4835.0001BAEA 571.38 CAD
H-4835.0001BA H-4835.0005BA
Human pTH (1-34) Acetate
Proteins and Peptides
See also the pTH2 receptor agonist TIP39 (H-4878).
Teriparatide, which consists of the N-terminal 34 amino acid residues of parathyroid hormone, shows the same potency and pharmacological profile as the native hormone. Administration of teriparatide stimulates bone formation and increases the bone mineral density (BMD).