(Asn⁶⁷⁰,Leu⁶⁷¹)-Amyloid beta/A4 Protein Precursor₇₇₀ (667-676) Trifluoroacetate
Supplier: BACHEM AMERICAS INC
4029475.0001 4029475.0005
H-4834.0001BAEA 167.67 CAD
H-4834.0001BA H-4834.0005BA
(Asn⁶⁷⁰,Leu⁶⁷¹)-Amyloid beta/A4 Protein Precursor₇₇₀ (667-676) Trifluoroacetate
Proteins and Peptides
This peptide substrate corresponds to the 'Swedish' Lys-Met/Asn-Leu (K670N/M671L) mutation of the amyloid precursor protein (APP) β-secretase cleavage site. It has been used for assaying β-secretase activity.