(Pyr³)-beta-Amyloid (3-42)
Supplier: BACHEM AMERICAS INC
4029424.0500 4029424.1000
H-4796.0500BAEA 1134.96 CAD
H-4796.0500BA H-4796.1000BA
(Pyr³)-beta-Amyloid (3-42)
Proteins and Peptides
(Pyr³)-Amyloid β-Protein (3-42) was found to be the predominant amyloid β-peptide structure deposited in human brain of Alzheimer's disease and Down's syndrome patients. Therefore, (Pyr³)-Aβ (3-42) is suggested to accumulate in the brain and to trigger the formation of insoluble amyloid β-peptide deposits. Nussbaum et al. studies the Prion-like behaviour and tau-dependent cytotoxicity of the truncated Aβ sequence.