Human Urotensin II Trifluoroacetate
Supplier: BACHEM AMERICAS INC
4029364.0001 4029364.0005
H-4768.0001BAEA 529.25 CAD
H-4768.0001BA H-4768.0005BA
Human Urotensin II Trifluoroacetate
Proteins and Peptides
Urotensin II is a vasoactive somatostatin-like cyclic peptide, which has long been regarded as a peptide exclusively produced by the urophysis of teleost fish. U-II has also been cloned from man. It was found to be the most potent mammalian vasoconstrictor identified so far. Furthermore, as urotensin II immunoreactivity was also found within central nervous system and endocrine tissues, it may have additional activities.