(D-Tyr⁵,D-Trp⁶)-LHRH Trifluoroacetate
Supplier: BACHEM AMERICAS INC
4025365.0001 4025365.0005
H-4646.0001BAEA 570.46 CAD
H-4646.0001BA H-4646.0005BA
(D-Tyr⁵,D-Trp⁶)-LHRH Trifluoroacetate
Proteins and Peptides
LHRH is also called Gonadotropin-Releasing Hormone (GnRH) or Luteinizing Hormone-Releasing Factor (LRF).
H-4075 corresponds to triptorelin acetate, H-6150 to triptorelin pamoate (triptorelin embonate).