(Des-Gly¹⁰,D-Tyr⁵,D-Leu⁶,Pro-NHEt⁹)-LHRH
Supplier: BACHEM AMERICAS INC
4028312.0001 4028312.0005
H-4638.0001BAEA 570.65 CAD
H-4638.0001BA H-4638.0005BA
(Des-Gly¹⁰,D-Tyr⁵,D-Leu⁶,Pro-NHEt⁹)-LHRH
Proteins and Peptides
LHRH is also called Gonadotropin-Releasing Hormone (GnRH) or Luteinizing Hormone-Releasing Factor (LRF).
Impurity G of the Leuprorelin Ph. Eur. monograph.