IPP
Supplier: BACHEM AMERICAS INC
4029177.0250
4029177.1000
H-4632.0250BAEA
1032.67
CAD
H-4632.0250BA
H-4632.1000BA
IPP
Proteins and Peptides
These short peptides act as angiotensin I-converting enzyme (ACE) inhibitors. See also Phe-Tyr (G-2955) and the subfamilies 'Bradykinin-Potentiating Peptide (BPP)' and 'Angiotensin I-Converting Enzyme (ACE, ACE2) Inhibitors'.
Antihypertensive tripeptide originally isolated from fermented milk. IPP inhibited angiotensin I-converting enzyme (ACE) with an IC₅₀ of 5 µM.