H-Phe-Phe-Phe-OH
Supplier: BACHEM AMERICAS INC
4011114.0250 4011114.1000
H-4560.0250BAEA 413.58 CAD
H-4560.0250BA H-4560.1000BA
H-Phe-Phe-Phe-OH
Proteins and Peptides
FFF is a bitter-tasting peptide due to its hydrophobicity. As diphenylalanine, triphenylalanine forms ordered nanostructures.