Rat Prolactin-Releasing Peptide (1-31)
Supplier: BACHEM AMERICAS INC
4028741.0500 4028741.1000
H-4384.0500BAEA 793.9 CAD
H-4384.0500BA H-4384.1000BA
Rat Prolactin-Releasing Peptide (1-31)
Proteins and Peptides
The hypothalamic releasing factors PrRP31 and PrRP20 which share no sequence similarity with known peptides and proteins, are neuropeptides from the brain that are synthesized as part of a larger precursor preprolactin. They have been found to promote the release of prolactin, the anterior pituitary hormone that is responsible for lactation.