H-Lys-Tyr-Lys-OH Acetate
Supplier: BACHEM AMERICAS INC
4000280.0250
4000280.1000
H-4280.0250BAEA
516.34
CAD
H-4280.0250BA
H-4280.1000BA
H-Lys-Tyr-Lys-OH Acetate
Proteins and Peptides
The tripeptide KYK is able to nick supercoiled or relaxed DNAs at apurinic/apyrimidinic sites. See also H-Lys-Trp-Lys-OH (H-4265).