LHRH (lamprey III) Trifluoroacetate
Supplier: BACHEM AMERICAS INC
4028403.0001 4028403.0005
H-4258.0001BAEA 258.14 CAD
H-4258.0001BA H-4258.0005BA
LHRH (lamprey III) Trifluoroacetate
Proteins and Peptides
LHRH is also called Gonadotropin-Releasing Hormone (GnRH) or Luteinizing Hormone-Releasing Factor (LRF).
Contrary to mammals and other classes of species, the sequences of GnRH I-III found in fish show a high diversity. H-4258, Lamprey GnRH III (peforelin) has found use in veterinary medicine and the sGnRH analog H-9205 in aquaculture.
Lamprey GnRH (also called peforelin) is a highly potent and specific FSH-releasing peptide that may enhance fertility in animals and humans. The peptide has found application in stimulating FSH release in swine. Peforelin surpasses GnRH in suppressing the growth of MDA-MB-231 and MCF-7 breast cancer cells. In comparison to other GnRH-III analogs, the peptide exhibits a superior antitumor activity.