(Des-Gly¹⁰,D-Trp⁶,Pro-NHEt⁹)-LHRH High Acetate
Supplier: BACHEM AMERICAS INC
4066809.0005 4066809.0025
H-4116.0005BAEA 180.91 CAD
H-4116.0005BA H-4116.0025BA
(Des-Gly¹⁰,D-Trp⁶,Pro-NHEt⁹)-LHRH High Acetate
Proteins and Peptides
LHRH is also called Gonadotropin-Releasing Hormone (GnRH) or Luteinizing Hormone-Releasing Factor (LRF).
Short-term administration of deslorelin stimulates the formation of LH and FSH, which induce an increase in the production of testosterone and estradiol. Prolonged administration of the GnRH agonist causes a sustained down-regulation of circulating LH and FSH levels and hence suppression of steroidogenesis.