Bovine;Human;Mouse Melanocyte Protein PMEL 17 (185-193) Trifluoroacetate
Supplier: BACHEM AMERICAS INC
4027973.0001 4027973.0005
H-4106.0001BAEA 464.74 CAD
H-4106.0001BA H-4106.0005BA
Bovine;Human;Mouse Melanocyte Protein PMEL 17 (185-193) Trifluoroacetate
Proteins and Peptides
For further melanoma-associated peptides please see also Fibronectin Fragments/RGD Peptides, Melanin-Concentrating Hormones (MCH) and Related Peptides, Peptide 74, Thrombospondin-1 (TS1) Cell Binding Domain (CBD) Fragments.
This peptide is as H-3958 a gp100 epitope that is recognized in the context of HLA-A2 by tumor-infiltrating lymphocytes (TIL) in vivo. PMEL 17 could be a melanogenic enzyme.