(Des-Gly¹⁰,D-Leu⁶,Pro-NHEt⁹)-LHRH Acetate, Bachem
Supplier: BACHEM AMERICAS INC
4033014.0005
4033014.0025
H-4060.0005BAEA
258.14
CAD
H-4060.0005BA
H-4060.0025BA
(Des-Gly¹⁰,D-Leu⁶,Pro-NHEt⁹)-LHRH Acetate, Bachem
Proteins and Peptides
LHRH is also called Gonadotropin-Releasing Hormone (GnRH) or Luteinizing Hormone-Releasing Factor (LRF).
Leuprolide, a highly active LHRH (GnRH) agonist stimulates the pituitary release of luteinizing hormone, causing an increase in testicular androgen production. Thus, leuprolide represents a safer alternative to estrogens, and may be preferred in the treatment of men with advanced prostate cancer. CAS Number (net): 53714-56-0.