Human beta-Endorphin (6-31)
Supplier: BACHEM AMERICAS INC
4030554.0001 4030554.0005
H-4024.0001BAEA 413.58 CAD
H-4024.0001BA H-4024.0005BA
Human beta-Endorphin (6-31)
Proteins and Peptides
For other opioid peptides see, the product families ‘Deltorphins and Dermorphins’, ‘Dynorphin, Analogs and Sequences, ‘Endorphins, MPF, Neoendorphins’, and ‘Opioid Peptides’.
This β-endorphin fragment, in which the C-terminus is preserved, functions as a non-opioid β-endorphin antagonist and mediates effects on the immune system.