H-Gly-Pen-Gly-Arg-Gly-Asp-Ser-Pro-Cys-Ala-OH Trifluoroacetate
Supplier: BACHEM AMERICAS INC
4027371.0001 4027371.0005
H-3964.0001BAEA 516.34 CAD
H-3964.0001BA H-3964.0005BA
H-Gly-Pen-Gly-Arg-Gly-Asp-Ser-Pro-Cys-Ala-OH Trifluoroacetate
Proteins and Peptides
GPenGRGDSPCA (cRGD), a cyclic RGD peptide has been used in the study of vascular wound response. GPenGRGDSPCA caused vasodilation when applied to isolated rat cremaster arterioles. The vasodilation is dependent on interaction of the soluble cRGD sequence with the αvβ3 integrin expressed by smooth muscle cells in the arteriolar wall.