Rat Procathepsin B (26-50)
Supplier: BACHEM AMERICAS INC
4028254.0500 4028254.1000
H-3942.0500BAEA 645.43 CAD
H-3942.0500BA H-3942.1000BA
Rat Procathepsin B (26-50)
Proteins and Peptides
Please see also the inhibitors H-7650, N-1000, N-1040, N-1105, N-1125, N-1315, N-1320, N-1380, N-1395, and N-1855.
This 25 residue peptide was completely resistant to proteolysis by cathepsin B even after incubation overnight. It inhibited human cathepsin B with Ki = 2.8 µM and rat cathepsin B with Ki = 2 µM.