H-Leu-Leu-Leu-OH
Supplier: BACHEM AMERICAS INC
4002312.0001
4002312.0005
H-3915.0001BAEA
396.79
CAD
H-3915.0001BA
H-3915.0005BA
H-Leu-Leu-Leu-OH
Proteins and Peptides
The tripeptide trileucine, found in casein and soy protein, was shown to increase the endothelial release of TXB₂, a stable marker of TXA₂. Inhibitor of human liver alanine aminopeptidase, Ki 0.054 mM. Addition of trileucine improved aerosol performance and stability of spray-dried powders for inhalation.