Human Tyrosinase (206-214) Acetate Salt
Supplier: BACHEM AMERICAS INC
4040740.0001 4040740.0005
H-3848.0001BAEA 264.44 CAD
H-3848.0001BA H-3848.0005BA
Human Tyrosinase (206-214) Acetate Salt
Enzymes
Sequence: H-Ala-Phe-Leu-Pro-Trp-His-Arg-Leu-Phe-OH
AFLPWHRLF is a tyrosinase epitope recognized by HLA-A24 restricted, tumor-infiltrating lymphocytes (TIL). Therefore it represents a reagent that may be used to generate melanoma-specific T cells for adoptive immunotherapy, as well as in peptide vaccines for HLA-A24+ melanoma patients.