H-D-Phe-Cys-Tyr-D-Trp-Arg-Thr-Pen-Thr-NH₂
Supplier: BACHEM AMERICAS INC
4027138.0001
4027138.0005
H-3698.0001BAEA
309.81
CAD
H-3698.0001BA
H-3698.0005BA
H-D-Phe-Cys-Tyr-D-Trp-Arg-Thr-Pen-Thr-NH₂
Proteins and Peptides
For other opioid peptides see, the product families ‘Deltorphins and Dermorphins’, ‘Endorphins, MPF, Neoendorphins’, and ‘Opioid Peptides'.
CTAP, an analog of CTOP, is a selective µ-opioid receptor antagonist.