H-Gly-Leu-Phe-OH
Supplier: BACHEM AMERICAS INC
4007924.0250 4007924.1000
H-3555.0250BAEA 570.65 CAD
H-3555.0250BA H-3555.1000BA
H-Gly-Leu-Phe-OH
Proteins and Peptides
See also the product families 'Adjuvant Peptides' and 'Thymosins and Fragments'.
Immunostimulating tripeptide from human and bovine α-lactalbumin. Administration of GLF prevented alopecia in neonatal rats treated with the anticancer agent etopside. See also H-Leu-Leu-Tyr-OH.