H-Cys-4-Abz-Met-OH
Supplier: BACHEM AMERICAS INC
4026861.0005
4026861.0025
H-3548.0005BAEA
309.81
CAD
H-3548.0005BA
H-3548.0025BA
H-Cys-4-Abz-Met-OH
Proteins and Peptides
Potent inhibitor of the Ras farnesyltransferase (Ftase) with an IC₅₀ of 50 nM. It prevented farnesylation of Ras and resulted in its inability to associate with other cell signaling components at the interior of the plasma membrane microenvironment.