H-Gly-Gly-Ile-OH
Supplier: BACHEM AMERICAS INC
4009599.0001 4009599.0005
H-3430.0001BAEA 516.34 CAD
H-3430.0001BA H-3430.0005BA
H-Gly-Gly-Ile-OH
Proteins and Peptides
The tripeptide GGI stimulates the Na⁺/Cl⁻-dependent transport system for opioid peptides in SK-N-SH cells, a human neuronal cell line.