Astressin Trifluoroacetate
Supplier: BACHEM AMERICAS INC
4026664.0500
4026664.1000
H-3422.0500BAEA
645.43
CAD
H-3422.0500BA
H-3422.1000BA
Astressin Trifluoroacetate
Proteins and Peptides
The Glu³⁰ to Lys³³ lactam bridge constrains astressin in an α-helical conformation which seems to be responsible for its especially high CRF antagonist activity. Astressin is 100 times more potent than α-helical CRF (9-41) (H-2040) at inhibiting ACTH secretion in vitro, and more than 10 times more potent in vivo than any other CRF antagonist reported to date.