H-Gly-Gly-His-OH
Supplier: BACHEM AMERICAS INC
4006117.0250
4006117.1000
H-3417.0250BAEA
206.51
CAD
H-3417.0250BA
H-3417.1000BA
H-Gly-Gly-His-OH
Proteins and Peptides
Short histidine-containing peptides can form stable complexes with copper and other transition metals. See also His-Leu (G-2310) and the product family 'Liver Cell Growth Factor (GHK) and Analogs'.
Cu(II) and Ni(II)-binding tripeptide.The Ni(II) complex of GGH was shown by Brown et al. to mediate protein crosslinking by oxidants. GGH has been used in biosensors for the electrochemical detection of Cu(II) ions.