Human;Rat Neuropeptide Y (3-36) Trifluoroacetate
Supplier: BACHEM AMERICAS INC
4031095.0500
4031095.1000
H-3326.0500BAEA
516.34
CAD
H-3326.0500BA
H-3326.1000BA
Human;Rat Neuropeptide Y (3-36) Trifluoroacetate
Proteins and Peptides
Main degradation product of NPY in serum and Y2/Y5 receptor agonist.