H-Gly-Gly-Arg-OH Acetate
Supplier: BACHEM AMERICAS INC
4003702.0250 4003702.1000
H-3305.0250BAEA 413.58 CAD
H-3305.0250BA H-3305.1000BA
H-Gly-Gly-Arg-OH Acetate
Proteins and Peptides
Mimic of mussel pheromones, induces the aggregation of various mussel species as Mytilis edulis. GGR also stimulates planktonic larvae to settle.