Human Osteostatin Trifluoroacetate, Bachem
Supplier: BACHEM AMERICAS INC
4031194.0500 4031194.1000
H-3212.0500BAEA 926.18 CAD
H-3212.0500BA H-3212.1000BA
Human Osteostatin Trifluoroacetate, Bachem
Proteins and Peptides
The carboxyl-terminal fragment of pTHrP, TRSAWLDSGVTGSGLEGDHLSDTSTTSLELDSR, is a potent inhibitor of osteoclastic bone resorption. It is suggested that this product acts directly on osteoclasts.