(Phenylac¹,D-Tyr(Et)²,Lys⁶,Arg⁸,des-Gly⁹)-Vasopressin Trifluoroacetate
Supplier: BACHEM AMERICAS INC
4031343.0001 4031343.0005
H-3186.0001BAEA 584.9 CAD
H-3186.0001BA H-3186.0005BA
(Phenylac¹,D-Tyr(Et)²,Lys⁶,Arg⁸,des-Gly⁹)-Vasopressin Trifluoroacetate
Proteins and Peptides
For desmopressin see H-7675.
Selective and highly potent V₁ receptor antagonist (antivasopressor) with an antiantidiuretic/antivasopressor ED ratio of about 570.