H-Gly-Arg-Gly-Asp-Asn-Pro-OH
Supplier: BACHEM AMERICAS INC
4030603.0005 4030603.0025
H-3174.0005BAEA 309.81 CAD
H-3174.0005BA H-3174.0025BA
H-Gly-Arg-Gly-Asp-Asn-Pro-OH
Proteins and Peptides
Potent inhibitor of cell attachment to fibronectin. GRGDNP promote cell adhesion preferentially via the α₅β₁ integrin.