H-Tyr-Arg-Gly-Asp-Ser-OH Trifluoroacetate
Supplier: BACHEM AMERICAS INC
4030618.0005 4030618.0025
H-3154.0005BAEA 423.95 CAD
H-3154.0005BA H-3154.0025BA
H-Tyr-Arg-Gly-Asp-Ser-OH Trifluoroacetate
Proteins and Peptides
PEG acrylate-based hydrogels have been modified with the RGD peptide YRGDS to improve their biocompatibility. YRGDS can be radioiodinated.