Human Growth Hormone (1-43)
Supplier: BACHEM AMERICAS INC
4030633.0500 4030633.1000
H-3146.0500BAEA 645.43 CAD
H-3146.0500BA H-3146.1000BA
Human Growth Hormone (1-43)
Proteins and Peptides
This insulin-potentiating hGH-fragment has been detected in human serum. Treatment of obese yellow Avy/A mice with hGH (1-43) enhanced the in vitro sensitivity of their adipose tissue to insulin.