Ophthalmic Acid
Supplier: BACHEM AMERICAS INC
4001847.0250
4001847.1000
H-3145.0250BAEA
929.4
CAD
H-3145.0250BA
H-3145.1000BA
Ophthalmic Acid
Proteins and Peptides
Acidic peptide of the lens. Carba analog of reduced glutathione. Ophthalmic acid could be used as a biomarker in oxidative stress.