Human (Leu¹⁵)-Gastrin I
Supplier: BACHEM AMERICAS INC
4011661.0001 4011661.0005
H-3090.0001BAEA 678.52 CAD
H-3090.0001BA H-3090.0005BA
Human (Leu¹⁵)-Gastrin I
Proteins and Peptides
This Gastrin I analog is as active as the native sequence (H-3085) but more stable. The activity loss of Gastrin due to Met sulfoxide formation is avoided.