Bovine PTH (3-34) Trifluoroacetate
Supplier: BACHEM AMERICAS INC
4031211.0500 4031211.1000
H-3088.0500BAEA 361.41 CAD
H-3088.0500BA H-3088.1000BA
Bovine PTH (3-34) Trifluoroacetate
Proteins and Peptides
See also the pTH2 receptor agonist TIP39 (H-4878).
In vitro experiments have shown that pTH (3-34) inhibits the stimulation of adenylate cyclase by pTH (1-34), but has no agonist or antagonistic effects on renal phosphate transport in vivo.