(D-Trp⁶)-LHRH (free acid) Trifluoroacetate
Supplier: BACHEM AMERICAS INC
4026526.0005 4026526.0025
H-3078.0005BAEA 258.14 CAD
H-3078.0005BA H-3078.0025BA
(D-Trp⁶)-LHRH (free acid) Trifluoroacetate
Proteins and Peptides
LHRH is also called Gonadotropin-Releasing Hormone (GnRH) or Luteinizing Hormone-Releasing Factor (LRF).
H-3078 may serve as a reference compound, as the free acid of triptorelin is a potential by-product or degradation product in pharmaceutical preparations of this GnRH agonist (H-4075).