(Ala¹·³·¹¹·¹⁵)-Endothelin-1 Trifluoroacetate, Bachem

Supplier: BACHEM AMERICAS INC

4030582.0500 4030582.1000
H-3066.0500BAEA 232.35 CAD
H-3066.0500BA H-3066.1000BA
(Ala¹·³·¹¹·¹⁵)-Endothelin-1 Trifluoroacetate, Bachem
Proteins and Peptides

Endothelins are peptides with exceptional vasoconstrictor potency. They play an important role in intercellular communications.


4 Ala ET-1, linear analog of ET-1 is a selective agonist for ET-B receptors.

Order Now

SPECIFICATIONS

Protein/peptide name (Ala¹·³·¹¹·¹⁵)-Endothelin-1 Trifluoroacetate
Environmentally Preferable
Species
Source
CAS No. 121204-87-3
MDL No. MFCD18633335
Sequence H-Ala-Ser-Ala-Ser-Ser-Leu-Met-Asp-Lys-Glu-Ala-Val-Tyr-Phe-Ala-His-Leu-Asp-Ile-Ile-Trp-OH
Molecular weight 2367.71
Molecular formula C₁₀₉H₁₆₃N₂₅O₃₂S
Storage conditions –20 ± 5 °C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR