beta-Amyloid (1-38) Trifluoroacetate
Supplier: BACHEM AMERICAS INC
4030204.0500 4030204.1000
H-2966.0500BAEA 528.73 CAD
H-2966.0500BA H-2966.1000BA
beta-Amyloid (1-38) Trifluoroacetate
Proteins and Peptides
Like Aβ (25-35) (H-1192), the Aβ fragment (1-38) destabilizes calcium homeostasis and renders human cortical neurones vulnerable to environmental insults.