beta-Amyloid (1-16) Trifluoroacetate

Supplier: BACHEM AMERICAS INC

4030330.0500 4030330.1000
H-2958.0500BAEA 176.52 CAD
H-2958.0500BA H-2958.1000BA
beta-Amyloid (1-16) Trifluoroacetate
Proteins and Peptides

The Cu²⁺ complex of this soluble amyloid β-protein fragment showed significant oxidative activities toward the catechol-like substrate 1,2,3-trihydroxylbenzene (pyrogallol) and plasmid DNA cleavage. The N-terminal Aβ fragments Aβ1-14 (H-7372), Aβ1-15 (H-6368), and Aβ1-16 are elevated in cell media and in CSF in response to γ-secretase inhibitor treatment. The fragments can serve as biomarkers in clinical studies of the effect of such inhibitors.

Order Now

SPECIFICATIONS

Protein/peptide name beta-Amyloid (1-16) Trifluoroacetate
Environmentally Preferable
Species
Source
CAS No. 131580-10-4
MDL No. MFCD18782174
Sequence H-Asp-Ala-Glu-Phe-Arg-His-Asp-Ser-Gly-Tyr-Glu-Val-His-His-Gln-Lys-OH
Molecular weight 1955.03
Molecular formula C₈₄H₁₁₉N₂₇O₂₈
Storage conditions –20 ± 5 °C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR