Human Adrenomedullin Trifluoroacetate
Supplier: BACHEM AMERICAS INC
4030315.0500
4030315.1000
H-2932.0500BAEA
1187.51
CAD
H-2932.0500BA
H-2932.1000BA
Human Adrenomedullin Trifluoroacetate
Proteins and Peptides
Adrenomedullin (ADM) elicits a potent and longlasting hypotensive effect and is considered as a hormone involved in blood pressure control. Additionally, ADM acts as a protective factor of blood vessels. Adrenomedullin is a potent stimulator of osteoblastic activity. Further biological activities of hADM include reduction of oxidative stress and inhibition of endothelial cell apoptosis.