Mating Factor alpha Trifluoroacetate
Supplier: BACHEM AMERICAS INC
4003514.0001 4003514.0005
H-2925.0001BAEA 219.73 CAD
H-2925.0001BA H-2925.0005BA
Mating Factor alpha Trifluoroacetate
Proteins and Peptides
Mating pheromone of mating type α cells of S. cerevisiae. This factor inhibits DNA synthesis, thereby preventing cell division of mating type a cells. The hydrophobic peptide binds to a class D G-protein-coupled receptor Ste2p.