(d(CH₂)₅¹,Tyr(Me)²,Thr⁴,Orn⁸,des-Gly-NH₂⁹)-Vasotocin Trifluoroacetate
Supplier: BACHEM AMERICAS INC
4031339.0001 4031339.0005
H-2908.0001BAEA 599.86 CAD
H-2908.0001BA H-2908.0005BA
(d(CH₂)₅¹,Tyr(Me)²,Thr⁴,Orn⁸,des-Gly-NH₂⁹)-Vasotocin Trifluoroacetate
Proteins and Peptides
For atosiban see H-6722.
OTA (dOVT), a potent and selective in vivo oxytocin antagonist.