(D-Pen²,D-Pen⁵)-Enkephalin Trifluoroacetate
Supplier: BACHEM AMERICAS INC
4008254.0001
4008254.0005
H-2905.0001BAEA
206.51
CAD
H-2905.0001BA
H-2905.0005BA
(D-Pen²,D-Pen⁵)-Enkephalin Trifluoroacetate
Proteins and Peptides
For other opioid peptides see, the product families ‘Deltorphins and Dermorphins’, ‘Endorphins, MPF, Neoendorphins’, and ‘Opioid Peptides'.
DPDPE mediates its direct antinociceptive actions via the δ₁-opioid receptor while its modulatory effects are mediated via the δ₂-opioid receptor.