(D-Ala⁷)-Angiotensin I/II (1-7) Trifluoroacetate
Supplier: BACHEM AMERICAS INC
4030357.0025 4030357.0100
H-2888.0025BAEA 949.93 CAD
H-2888.0025BA H-2888.0100BA
(D-Ala⁷)-Angiotensin I/II (1-7) Trifluoroacetate
Proteins and Peptides
The sequences given for angiotensin I (H-1680) and II (H-1705) correspond to the sequences of human, horse, sheep, pig, mouse, and rat angiotensins I and II.
A-779 is a potent and selective angiotensin (1-7) antagonist.