H-Tyr-Ile-Gly-Ser-Arg-NH₂
Supplier: BACHEM AMERICAS INC
4030799.0005 4030799.0025
H-2802.0005BAEA 413.58 CAD
H-2802.0005BA H-2802.0025BA
H-Tyr-Ile-Gly-Ser-Arg-NH₂
Proteins and Peptides
YIGSR-amide binds to the laminin receptor and inhibits experimental metastasis formation. In comparison to YIGSR (H-6825), C-terminal amidation increases the activity of the peptide significantly.