Human;Mouse H-Cys-Asp-Pro-Gly-Tyr-Ile-Gly-Ser-Arg-OH Trifluoroacetate
Supplier: BACHEM AMERICAS INC
4030804.0001 4030804.0005
H-2798.0001BAEA 185.3 CAD
H-2798.0001BA H-2798.0005BA
Human;Mouse H-Cys-Asp-Pro-Gly-Tyr-Ile-Gly-Ser-Arg-OH Trifluoroacetate
Proteins and Peptides
CDPGYIGSR corresponds to residues 925-933 of the laminin B1 chain. Like the peptide amide of the same sequence (H-1224), it also binds to the laminin receptor, but with lower affinity.