Dog;Human;Pig (Deamino-Phe¹⁹,D-Ala²⁴,D-Pro²⁶-psi(CH₂NH)Phe²⁷)-GRP (19-27) Trifluoroacetate
Supplier: BACHEM AMERICAS INC
4030653.0500 4030653.1000
H-2756.0500BAEA 800.3 CAD
H-2756.0500BA H-2756.1000BA
Dog;Human;Pig (Deamino-Phe¹⁹,D-Ala²⁴,D-Pro²⁶-psi(CH₂NH)Phe²⁷)-GRP (19-27) Trifluoroacetate
Proteins and Peptides
BW10 (or BW2258U89, Deamino-Phe-His-Trp-Ala-Val-D-Ala-His-D-Pro- psi(CH₂NH)-Phe-NH₂), a bombesin receptor antagonist, potently inhibits bombesin evoked release of gastrointestinal hormones from rats and dogs, in vitro and in vivo