µ-Conotoxin GIIIA
Supplier: BACHEM AMERICAS INC
4030525.0500 4030525.1000
H-2738.0500BAEA 1549.03 CAD
H-2738.0500BA H-2738.1000BA
µ-Conotoxin GIIIA
Proteins and Peptides
µ-Conotoxin GIIIA from the Conus geographus L. venom blocks with very high selectivity the muscle subtype of sodium channels.