Human Dynorphin A Trifluoroacetate
Supplier: BACHEM AMERICAS INC
4007832.0001 4007832.0005
H-2620.0001BAEA 464.74 CAD
H-2620.0001BA H-2620.0005BA
Human Dynorphin A Trifluoroacetate
Proteins and Peptides
The N-terminus of dynorphin, YGGFL, corresponds to Leu-enkephalin. So, dynorphin A (1-5) amide is filed as Leu-enkephalin amide (H-2745). For other opioid peptides see, the product families ‘Deltorphins and Dermorphins’, ‘Endorphins, MPF, Neoendorphins’, and ‘Opioid Peptides'.
Endogenous kappa-receptor antagonist.